- Home
- Incubators
- Cooled Incubators
- Cooled incubators with Peltier, IPP/IPPplus
Cooled incubators with Peltier, IPP/IPPplus
Supplier: Memmert
390-0621EA
6038
EUR
390-0621
390-0271
Cooled incubators with Peltier, IPP/IPPplus
Incubators
Cooled Incubators
The range has a choice of Peltier cooled models with a temperature range up from zero up to 70 °C. There is a choice of two different displays, SingleDISPLAY or TwinDISPLAY depending on the application. Peltier technology provides energy efficient cooling and heating. The Peltier elements can be controlled individually ensuring homogenous temperature and humidity distribution in the chamber. All models have stainless steel inner chamber and housing, ensuring long working life and easy cleaning. These cooled incubators have an integrated data logger and digital timer which is adjustable from one minute to 99 days and 23 hours.
- SingleDISPLAY IPP models, ideal for standard applications:
- ControlCOCKPIT parameters: Temperature (Celsius or Fahrenheit), program time, time zones and daylight-saving time
- One PT100 temperature sensor in a 4-wire circuit and ethernet interface for reading out the protocol log
- Adjustable overtemperature monitor and mechanical temperature limiter
IPPplus models with TwinDISPLAY feature the intuitive control and logging software, AtmoCONTROL, which uses drag and drop symbols to input values. The software allows online monitoring of up to 32 connected appliances and can send an automatic alarm message to one or several e-mail addresses. Complex ramping processes can be programmed quickly and easily for TwinDISPLAY models. The SetpointWAIT function ensures the process time does not start before the set temperature is reached. This feature can also be used with additional, freely positionable PT100 sensors, so that the process time starts when the set temperature is reached at all measuring points.
TwinDISPLAY IPPplus models, ideal for complex ramping processes:
*Intuitive, user friendly ovens with 2 TFT displays. ControlCOCKPIT with USB port. ControlCOCKPIT parameters as above
*Two PT100 sensors in a 4-wire circuit for mutual monitoring, taking over functions in case of an error
*The HeatBALANCE function allows application specific adjustment of heating/cooling power distribution (balance) - upper and lower Peltier elements for heating/cooling
*Logged protocol data is displayed on the ControlCOCKPIT (max 10 000 values correspond to approx. 1 week)
*Ethernet interface for reading out the protocol log and for uploading and implementing programs and for online logging
Multiple overtemperature protection with audible and visual alarm: Electronic temperature monitoring TWW safety device Class 3.3 or TWB safety device Class 2, adjustable on display (independent adjustable over-temperature which protects the samples and the chamber) and mechanical temperature limiter TB (protection Class 1) according to DIN 12880. AutoSAFETY automatically adjusts to the set value within a freely adjustable tolerance range. Setting individual min/max values for over- or under-temperature.
*Two PT100 sensors in a 4-wire circuit for mutual monitoring, taking over functions in case of an error
*The HeatBALANCE function allows application specific adjustment of heating/cooling power distribution (balance) - upper and lower Peltier elements for heating/cooling
*Logged protocol data is displayed on the ControlCOCKPIT (max 10 000 values correspond to approx. 1 week)
*Ethernet interface for reading out the protocol log and for uploading and implementing programs and for online logging
Multiple overtemperature protection with audible and visual alarm: Electronic temperature monitoring TWW safety device Class 3.3 or TWB safety device Class 2, adjustable on display (independent adjustable over-temperature which protects the samples and the chamber) and mechanical temperature limiter TB (protection Class 1) according to DIN 12880. AutoSAFETY automatically adjusts to the set value within a freely adjustable tolerance range. Setting individual min/max values for over- or under-temperature.
Accessories information: For details of the range of shelves and grids, please visit website or contact your local Avantor sales office.
Ordering information: Supplied without grids or shelves, please order required shelves, stainless steel grids or trays separately. Range of accessories and factory fitted options such as access ports, interior sockets and lighting are available on request. Please contact Avantor for further details prior to ordering as these can not be retro-fitted.
New Product
Limited Time Sale
Select items below are currently on sale
Services: Memmert IPP/IPPplus cooled incubators have a 3 year warranty period as standard. This is an additional 1 year extension to that 3 year period, giving 4 years in total.
Enter Certificate Details
Inquire for Price
Learn more
About VWR
Item(s) Added to Cart