Pipette controller, Powerpette
613-4449EA 291 EUR
JENC266-069 JENC266-082 JENC266-070 JENC266-081 JENC266-008 JENC266-007 JENC266-086 JENC266-074 JENC266-085 JENC266-073 JENC266-078 JENC266-077
Pipette controller, Powerpette
Pipette controller for glass and plastic pipettes from 1 to 100 ml.
  • Finger triggers to minimize any high pressure points
  • Mode selection switch enables selection of low or high modes, both with variable aspirate and dispense speeds
  • Operate on two rechargeable, long-life batteries
These lightweight, cordless pipetting aids offer precision fingertip control in a unique streamlined design.
Delivery information: Supplied with 0,45 µl and 0,2 µl hydrophobic filters, charger, bench stand, wall stand and instruction manual.