VWR® Bags with Fast-Zip Closure and Pocket for Documents
Supplier: Avantor
VWR®
720-1978EA 82.6 EUR
720-1978
VWR® Bags with Fast-Zip Closure and Pocket for Documents
Bags Sample Bags
Transparent polythene bags for biological transport with English instructions, plus a pocket for documents and printed biohazard logo.
  • Fast-zip closure for easy opening and closing without risk of leakage
  • Double-pocket bag: Two separate environments for laboratory documents and specimens
  • The sample storage condition legend is printed on the bag (frozen, refrigerated, room temperature)