You Searched For: Stain Removal Reagents
7  results were found
SearchResultCount:"7"
Sort Results
List View Easy View (new)
Rate These Search Results
Catalog Number: (E732-25)
Supplier: VWR Chemicals
Description: Destaining bags are designed to safely remove ethidium bromide from solution during overnight treatment. Each bag extracts up to 5 mg of ethidium brom...
UOM: 1 * 25 items
Catalog Number: (22007.)
Supplier: Biotium
Description: Activated charcoal retains ethidium bromide, GelRedâ„¢ and GelGreenâ„¢ efficiently in a convenient bag for decontamination of used gel staining solutions....
UOM: 1 * 25 items
MSDS
Catalog Number: (786-199)
Supplier: G-Biosciences
Description: The staining of gels with silver stains often results in over-staining or unevenly stained gels. Silver D-Stainâ„¢ eliminates the need to repeat the ele...
UOM: 1 * 25 items
Supplier: G-Biosciences
Description: G-Biosciences' Destain I contains approximately 45% methanol and 9% glacial acetic acid. G-Biosciences' Destain II contains approximately 5% methanol ...
Catalog Number: (ICNA112350200)
Supplier: MP Biomedicals
Description: The EtBr Greenbagâ„¢ Disposal Kit allows rapid and trouble-free concentration of ethdium bromide from large volumes of solutions into a small 'teabag' c...
UOM: 1 * 50 items
MSDS
Catalog Number: (J62931.8U)
Supplier: Thermo Scientific
Description: Ethidium bromide destaining bags are used to safely remove ethidium bromide from nucleotide solutions and gels during overnight treatment. Each bag ex...
UOM: 1 * 25 items
Catalog Number: (786-499)
Supplier: G-Biosciences
Description: After Coomassie Brilliant Blue staining process, the band intensity may be further enhanced by de-staining the stained gel in our CBB De-Staining Solu...
UOM: 1 * 1 L
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 7 of 7
no targeter for Bottom