VWR® Traceable®, Digital Thermo-Hygrometer
Supplier: VWR
VWR® Traceable®
620-1921EA 71.3 EUR
620-1921 VWRU35519-047 VWRU35519-046
VWR® Traceable®, Digital Thermo-Hygrometer
Hygrometers Digital Thermo-Hygrometers
This thermo-hygrometer is suitable for routine measurements, monitoring round the clock, quality controls and critical experimental requirements. It shows the exact air conditions in hoods, cleanrooms, storage rooms, environmental chambers and chemicals storage areas.
  • Continuous and simultaneous display of relative humidity and temperature
  • Minimum/maximum memories
  • Fast response
W×D×H: 57×85×6 mm
Weight: 70 g
Conforms to ISO/IEC 17025, ANSI/NCSL Z540-1 and 9001 accredited by A2LA. Traceable to NIST.
Certifications: Conforms to ISO/IEC 17025, ANSI/NCSL Z540-1 and 9001 accredited by A2LA. Traceable to NIST.
Delivery information: Supplied with Traceable® certificate of calibration, wall mounting and bench stand, solid sensors plus two AAA batteries.
RH = Relative Humidity