Portable pivoting davit column, MN50-POTEX
Supplier: Miller by Honeywell
DALL1002920EA 3130 EUR
DALL1002920
Portable pivoting davit column, MN50-POTEX
Max working load: 165 kg
  • Integrated rescue winch (30 m)
  • Adjustable legs to fit most confine space entry