Magnetic hotplate stirrer, SLHS
Supplier: SI Analytics
SCOI285416410EA 538 EUR
SCOI285416410 SCOI285416400
Magnetic hotplate stirrer, SLHS
Stirrers Magnetic Hotplate Stirrers
These ceramic coated stainless steel hot plate stirrers are compatible with most standard size borosilicate bottles, beakers and flasks. The compact design allows them to be used on crowded laboratory benches or under fume hoods.
  • Highly resistant to strong acids and bases
  • Electronic control to prevent overcharging
  • Two control LEDs to indicate heating and stirring function are on
The powder coated aluminium cast housing helps to resist heat. The drive motor is equipped with ball bearings for smooth running and a long service life.