Vacuum gel drying system, GD2000
Supplier: Hoefer
HOEFGD2000-230VEA 6312 EUR
HOEFGD2000-230V HOEFGD2002
Vacuum gel drying system, GD2000
The GD2000 vacuum gel drying system uses a dry heat vacuum method to dry gels rapidly, evenly, and safely.
  • Dries acrylamide and agarose gels as large as 33×44 cm in as little as 30 minutes
  • Heat and vacuum are applied from beneath the gel through a heavy aluminium platen for even heat and vacuum distribution under the gel
  • Independent timers for heat and for vacuum
  • Thermostat adjusts from 40 to 80 ±2 °C
  • A quiet, oil-free diaphragm pump maintains the low vacuum necessary for rapid gel drying
Delivery information: Complete system includes GD2000 gel dryer with silicone rubber overlay sheet, VP200 vacuum pump, stainless steel screen, porous PE cover sheet, Mylar® cover sheet, 50× porous cellophane sheets, 10× filter paper, and 3 m vacuum tubing. Basic unit is supplied without VP200 vacuum pump or vacuum tubing.