You Searched For: Micro Valves
18  results were found
SearchResultCount:"18"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: ENTEGRIS INC
Description: Available compliant to SEMI Standard F57-0312, the specification for polymer components used in ultrapure water and liquid chemical distribution syste...
Catalog Number: (ENTED8-3Q-8F)
Supplier: ENTEGRIS INC
Description: Available compliant to SEMI Standard F57-0312, the specification for polymer components used in ultrapure water and liquid chemical distribution syste...
UOM: 1 * 1 items
Supplier: ENTEGRIS INC
Description: CR4 pneumatic valves are specially designed to handle temperatures upto 160 °C. There is a option to add ¼" and ¹/₈" connections from Flaretek®. With ...
Supplier: ENTEGRIS INC
Description: O-ring free design, ¹/₂ or ¹/₄" orifice.
Catalog Number: (ENTE201-42-01-3)
Supplier: ENTEGRIS INC
Description: Available compliant to SEMI Standard F57-0312, the specification for polymer components used in ultrapure water and liquid chemical distribution syste...
UOM: 1 * 1 items
Supplier: ENTEGRIS INC
Description: This 3-way valve (patent-pending) has an all-PFA valve body and high purity fluoropolymer wetted components providing a noncontaminating fluid stream ...
Supplier: ENTEGRIS INC
Description: This 3-way valve (patent-pending) has an all-PFA valve body and high purity fluoropolymer wetted components providing a noncontaminating fluid stream ...
Supplier: ENTEGRIS INC
Description: Smaller than most comparable valves, ¹/₄" CR4 series valves are ideal for handling wet etch and clean process chemicals. These durable valves can hand...
Supplier: ENTEGRIS INC
Description: Available compliant to SEMI Standard F57-0312, the specification for polymer components used in ultrapure water and liquid chemical distribution syste...
Supplier: ENTEGRIS INC
Description: Available compliant to SEMI Standard F57-0312, the specification for polymer components used in ultrapure water and liquid chemical distribution syste...
Catalog Number: (ENTE202-86-01)
Supplier: ENTEGRIS INC
Description: Available compliant to SEMI Standard F57-0312, the specification for polymer components used in ultrapure water and liquid chemical distribution syste...
UOM: 1 * 1 items
Catalog Number: (ENTE202-82-01)
Supplier: ENTEGRIS INC
Description: Available compliant to SEMI Standard F57-0312, the specification for polymer components used in ultrapure water and liquid chemical distribution syste...
UOM: 1 * 1 items
Catalog Number: (ENTE202-128-01)
Supplier: ENTEGRIS INC
Description: Available compliant to SEMI Standard F57-0312, the specification for polymer components used in ultrapure water and liquid chemical distribution syste...
UOM: 1 * 1 items
Supplier: ENTEGRIS INC
Description: Available compliant to SEMI Standard F57-0312, the specification for polymer components used in ultrapure water and liquid chemical distribution syste...
Supplier: HAMILTON BONADUZ
Description: HV plug valves provide an economical and flexible method of custom-building a fluid flow control system, from a simple on/off valve to a more complex ...
Supplier: JOUR RESEARCH (VICI)
Description: These micro valves for LC and GC has a unique design of the fitting detail which allows a leak-free seal with no potential for rotor damage from overt...
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 18
no targeter for Bottom