Hollow fiber dialysis modules, Spectra/Por®
Supplier: Spectrum
SPEC500-016EA 383 EUR
SPEC500-016
Hollow fiber dialysis modules, Spectra/Por®
Hollow fiber dialysis modules offers a highly efficient, counter-current dialysis device convenient and easy enough to operate in the laboratory.
  • Hollow fiber membrane geometry
  • Inlet and outlet flow-stream connection ports
  • Achieve higher purities
  • Dialysis of 100 ml to multi-liter samples