Beta radiation shields, personal carrrier
Supplier: CBS Scientific
CBSCRL-024BEA 293 EUR
CBSCRL-024B
Beta radiation shields, personal carrrier
Acrylic carrier shield, transparent, thickness: 12,7 mm.
  • Carrier shield with 24 places for 1,5 to 2,0 ml Eppendorf tubes
  • Features hinged lid and an ice trough to keep samples cool
  • Permits mobility and protection without having to use a designated radioactive work station