CO₂ incubators, INCOmed series
Supplier: Memmert
390-0674EA
6229
EUR
MEMMINC246MED
MEMMINC153MED
CO₂ incubators, INCOmed series
These three standard units can be upgraded with six additional modules for optimal hygiene, comfort, documentation and programming. The Comfort module has two gas connections with quick release connectors and an automatic switch-over of gas cylinders. The electropolished interior of the Hygiene module is seamlessly welded by laser to ensure minimum contamination.
- Uniform atmosphere and temperature distribution due to enclosed, non-turbulent ventilation system in chamber
- Hot air sterilisation at 160 °C, four hour program
- Low contamination risk in the seamless, deep-drawn stainless steel inner chamber, which has additional door and rear heating to avoid condensation
- Multifunctional, digital microprocessor PID-controller
- Two Class A PT100 sensors, independently monitoring and taking over the performance at the same temperature value
The Communication module includes USB interface, “Celsius“ standard software for programming and logging, ring log memory and printer interface. The CO₂ module has an extended CO₂ range from 0 to 20%. The Premium module includes the Comfort, Hygiene, Communication and CO₂ modules. The Humidity module enables active microprocesssor controlled humidification and dehumidification (40 - 97% RH). All models have a fully insulated stainless steel door with double locking and four point adjustment and an inner glass door with opening (8 mm Ø) to take gas samples. Units have microprocessor control for temperature and CO₂ with various alarm and status displays, built in water condensation control. Digital (LED) display for all set parameters and digital seven-day program timer with real time clock. All include various safety features including an independent adjustable temperature safety device Class 3.1 with a visual alarm, saves the samples and the incubator, audible and visual alarm in case of over- or under-temperature, over- or under- CO₂, open door and empty gas cylinder.
Ordering information: Supplied without grids or water dish, please order required stainless steel grids, perforated shelves or reinforced grids separately, details of grids can be found at vwr.com. Range of accessories and factory fitted options such as LED light modules, access ports and interior sockets are available on request. Please contact VWR for further details prior to ordering as these cannot be retro-fitted. Fresh water supply module: To be used in combination with humidity module only. Memmert INCOmed CO₂ incubators have a 3 year warranty period as standard. There is an optional additional 1 year extension to that 3 year period, giving 4 years in total.
Caution: For details of the range of shelves and grids, please visit vwr.com or contact your local VWR sales office.
New Product
Limited Time Sale
Select items below are currently on sale
Enter Certificate Details
Item(s) Added to Cart