Inoculating needles, Sterilin®
Supplier: Thermo Scientific
SLINSN20EA 172 EUR
SLINSN20
Inoculating needles, Sterilin®
Sterilin® inoculating needles are ideal for picking off individual colonies within mixed cultures growing on plated media. They are also suitable for making stab inoculations into agar slants or tubes of solid culture media.