E-64
J586-5MGEA 0 EUR
VWRCJ586-5MG
E-64
An irreversible inhibitor of cysteine proteases like papain, calpain, and cathepsin B, H, L and S. Also an effective inhibitor of collegenase.
  • Purity (HPLC)- ≥98%