VWR®, Pig Trypsin (from Pancreas)
Supplier: VWR
0458-250GEA 0 EUR
VWRC0458-250G
VWR®, Pig Trypsin (from Pancreas)
Trypsin from porcine pancreas used in special preparation of single cell suspension.
EINECS: 232-650-8
GHS Symbol 1: GHS08
GHS Symbol 2: GHS07
MDL: MFCD00082094