You Searched For: Fall Protection Kits
4  results were found
SearchResultCount:"4"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: CAPITAL SAFETY DEUTSCHLAND
Description: Protecta® Cabloc® Vertical Lifeline System consists of an 8 mm cable which is terminated at the top by a cable ferrule and cable clamps at the bottom.
Supplier: CAPITAL SAFETY DEUTSCHLAND
Description: The Rollgliss R350 system provides efficient hauling, but more importantly, very controlled lowering (a finger and thumb are all that is required to h...
Catalog Number: (139-0234)
Supplier: HONEYWELL SAFETY
Description: Safety kit for maintenance work at industrial plants, tasks on antennae and construction sites.
UOM: 1 * 1 items
Catalog Number: (113-1271)
Supplier: HONEYWELL SAFETY
Description: Fall protection kit contains:
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 4 of 4
no targeter for Bottom