Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "office-furniture"
https://ru-bot.vwr.com/stibo/smallweb/std.lang.all/96/03/18989603.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"19"
  List View Grid View Easy View (new)
Sort by:
Supplier:  HAMMERBACHER
Description:   The table top is 25 mm thick, made of walnut, melamine coated according to DIN 68765. The C-shaped frame is made of round tubes with a diameter of 35 ...
Supplier:  HAMMERBACHER
Description:   Can be used individually, large variety of colours. Carcass construction, i.e. each rack can stand on its own.
Supplier:  HAMMERBACHER
Description:   Pedestal AC30 SG Grey 1 * 1 items
Supplier:  HAMMERBACHER
Description:   Pedestal TC30 CE Maple 1 * 1 items
Supplier:  HAMMERBACHER
Description:   Desk series with a range of variation and combination options.
Supplier:  HAMMERBACHER
Description:   Pedestal TC20 WE Maple 1 * 1 items
Supplier:  HAMMERBACHER
Description:   Standcontainer SC50 WE Maple 1 * 1 items
Supplier:  HAMMERBACHER
Description:   Pedestal AC20 BM Plum/White 1 * 1 items
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 8  of 19
  1  2  3  Next