You Searched For: Drum Storage
38  results were found
SearchResultCount:"38"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: asecos
Description: This sump pallets are made of PE with varied capacities from 60 to 400 L.
Supplier: asecos
Description: Available either painted (RAL 5010Gentian blue) or hot dip galvanised.
Supplier: asecos
Description: Blue polyethylene sump pallet with 1100 L capacity for the storage of IBC.
Supplier: asecos
Description: Polyethylene sumppallet with 1000 litres capacity for the storage of IBC.
Supplier: asecos
Description: Steel, transportable.
Supplier: asecos
Description: Painted or galvanised steel.
Catalog Number: (NEWPDLY301-01)
Supplier: NEW PIG
Description: This drum dolly with adjustable diameter quickly expands for a precise fit on drums or overpacks up to Ø 677 mm and 363 kg.
UOM: 1 * 1 items
Supplier: NEW PIG
Description: These spill pallets are ideal for long-term storage of liquids with high specific gravities and molded-in sump catches leaks, drips and spills.
Supplier: CEMO GMBH
Description: These small container/laboratory trays can be combined to cover a complete storage area and also can be used as storage/filling station. Trays can be ...
Supplier: CEMO GMBH
Description: Drum racks for approved and space saving storage of barrels and other containers.
Supplier: CEMO GMBH
Description: The steel sumps provide safe storage for water-polluting as well as inflammable substances.
Supplier: JUSTRITE MANUFACTURING CO
Description: A robust design made of 55% recycled polyethylene features convenient rolltop doors to allow access from front or rear.
Supplier: CEMO GMBH
Description: These sump pallets offer protection and a clean environment, through high resistance against acids and lyes and are used for the storage of caustic, f...
Supplier: CEMO GMBH
Description: These sumps are manufactured from high quality polyethylene (HD-LMD).
Catalog Number: (NEWPPAK601-AWO)
Supplier: NEW PIG
Description: These roll top hardcovers can be used for all-weather protection and liquid containment. Durable swing-out doors on front offers easy loading and unlo...
UOM: 1 * 1 items
Supplier: CEMO GMBH
Description: The sumps are applicable for the storage of environmentally hazardous substances.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 38
no targeter for Bottom