You Searched For: Kjeldahl Tablets and Reagents
5  results were found
SearchResultCount:"5"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Chemicals
Description: Kjeldahl tablets are catalysts that promote the digestion step of the Kjeldahl method for nitrogen determination.
Catalog Number: (330666A)
Supplier: VWR Chemicals
Description: This product is not available in all countries. Please check with your local Avantor International office or supplier.
UOM: 1 * 1.000 Tablet
Catalog Number: (ACRO243920025)
Supplier: Thermo Scientific
Description: Kjeldahl tablets with selenium (Wieninger), 5 g, Contents: Na₂SO₄: 4875 mg, Cu(II)SO₄.5H₂O: 75 mg; Se: 50 mg, Ø: 18 - 20 mm
UOM: 1 * 2,5 kg
MSDS
Supplier: THOMPSON & CAPPER
Description: Kjeldahl catalyst reagents in a convenient tablet form with a range of tablets to suit general and specific applications.
Catalog Number: (GERH12-0326)
Supplier: GERHARDT
Description: Kjeltab ST, 3,5 g, Contents: K2SO4 3,5 g; Se 0,0035 g, Ø: 16 mm
UOM: 1 * 1.000 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 5 of 5
no targeter for Bottom