You Searched For: PCR Tests
29  results were found
SearchResultCount:"29"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: BIOPREMIER
Description: Food safe kits provide a simple, reliable, and rapid procedure for detecting the presence of an animal species in meat and meat products. The assay is...
Supplier: BIOPREMIER
Description: PCR Tests, Real Time Detection Kit Lupin
Supplier: BIOPREMIER
Description: Food safe kits provide a simple, reliable, and rapid procedure for detecting the presence of a specific pathogenic bacteria. The assay is based on 5’ ...
Supplier: PRIMERDESIGN
Description: PCR kits for the detection and simultaneous quantification of human herpes viruses.
Supplier: PRIMERDESIGN
Description: PCR kits for the detection and simultaneous quantification of gastrointestinal pathogens.
Supplier: PRIMERDESIGN
Description: Papillomaviruses are a diverse group of DNA-based viruses that infect the skin and mucous membranes of humans and a variety of animals. Although more ...
Supplier: PRIMERDESIGN
Description: PCR kits for the detection and simultaneous quantification of hepatitis viruses.
Catalog Number: (SARTSMB95-1003)
Supplier: Sartorius
Description: The Microsart® ATMP Mycoplasma detection kit is especially designed for an easy and fast detection of mycoplasma in cell-based therapeutics like autol...
UOM: 1 * 1 items
Supplier: Sartorius
Description: The Microsart® RESEARCH Mycoplasma real-time PCR kit is especially designed for fast and reliable Mycoplasma contamination control in cell cultures, c...
Supplier: Q-BIOANALYTIC GMBH
Description: Pseudomonas aeruginosa is tested usually with cultural methods. However, this may take several days to deliver results. Using this Real-Time PCR test...
Supplier: Q-BIOANALYTIC GMBH
Description: Vibrio alginolyticus causes eye, ear and wound infections. It can be tested using this Real-Time PCR test. The analysis can be done in water and food ...
Supplier: Q-BIOANALYTIC GMBH
Description: This product range is optimised for real-Time PCR and uses Taqman® probe technology for the detection of human pathogenic bacteria. The kits are typic...
Supplier: Q-BIOANALYTIC GMBH
Description: OneCup it‘s so easy! The OneCup assay comes along with a minimum of pipetting steps. Everything is included in the mix. You only need to combine 20μl ...
Supplier: Q-BIOANALYTIC GMBH
Description: Successful PCR applications depend on well purified DNA. In diagnostics and analytics of food and medical samples PCR and real-time PCR has many advan...
Supplier: Q-BIOANALYTIC GMBH
Description: Kit for Vibrio cholerae tox gene (ctx). Food safety experts like the German DGHM recommend to test warm water fish for vibrio cholerae. With this Real...
Supplier: Q-BIOANALYTIC GMBH
Description: Vibrio vulnificus causes severe wound infections and is harmful when ingested. 33% of patients being infected die. The detection in food and water sam...
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 29
no targeter for Bottom