You Searched For: Microbiology Test Systems
99  results were found
SearchResultCount:"99"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Chemicals
Description: Confirmative test for coagulase-positive Staphylococcus.
Supplier: VWR Chemicals
Description: Microbiological laboratories need a simple procedure to maintain important microorganisms rather than repetitive sub-culturing, which results in conta...
Catalog Number: (941970ZA)
Supplier: VWR Chemicals
Description: The catalase test can differentiate or identify microorganisms by checking their ability to produce catalase, an enzyme that protects microorganisms f...
UOM: 1 * 30 mL
Catalog Number: (942152ZF)
Supplier: VWR Chemicals
Description: Swabs imprenated with reagent for the detection and verification of bacterial cytochrome oxidase.
UOM: 1 * 50 Tests
Supplier: THERMO FISHER DIAGNOSTICS GMBH
Description: Manually determine the antibiotic susceptibility of microorganisms using Antimicrobial Susceptibility Testing (AST) methods in conjunction with reliab...
Supplier: THERMO FISHER DIAGNOSTICS GMBH
Description: RapIDâ„¢ Systems are a complete line of cost-effective manual identification panels for a broad spectrum of microorganisms. RapIDâ„¢ panels offer easy, on...
Supplier: THERMO FISHER DIAGNOSTICS GMBH
Description: Enzymatic Tests, Microbactâ„¢ Reagent VP I
Supplier: THERMO FISHER DIAGNOSTICS GMBH
Description: Remel Agglutinating Sera are used for serological identification of Bordetella cultures by rapid slide agglutination test.
Supplier: LIOFILCHEM S.R.L.
Description: EnteroPluri Test is a 12-sector system containing special culture media that permits identification of the Enterobacteriaceae and other gram ne...
Supplier: LIOFILCHEM S.R.L.
Description: Antibiotic Tests, Chloramphenicol C 30 µg
Supplier: THERMO FISHER DIAGNOSTICS GMBH
Description: 25 discs/vial.
Supplier: LIOFILCHEM S.R.L.
Description: Lyophilised reference stock culture preparations containing a single strain of a microorganism for quality control of culture media, education and pro...
Supplier: THERMO FISHER DIAGNOSTICS GMBH
Description: The Oxoid Biochemical Identification System (O.B.I.S.) is a rapid colorimetric test for the determination of biochemical activity in microorganisms. I...
Supplier: FUJIFILM WAKO CHEMICALS
Description: The PYROSTARâ„¢ ES-F series of reagents represent our uniquely endotoxin specific line of Limulus Amebocyte Lysate products, which are formulated to be ...
New Product
Supplier: FUJIFILM WAKO CHEMICALS
Description: The PYROSTARâ„¢ ES-F series of reagents represent our uniquely endotoxin specific line of Limulus Amebocyte Lysate products, which are formulated to be ...
New Product
Catalog Number: (A9019.0025)
Supplier: PanReac AppliChem
Description: Lyophilised PCR mix for the detection of mycoplasma in cell culture by Real-Time/qPCR.
UOM: 1 * 25 Tests
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 99
no targeter for Bottom