You Searched For: Turbidity Standards
18  results were found
SearchResultCount:"18"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Chemicals
Description: All turbidity standard solutions are Certified Reference Materials.
Catalog Number: (84209.600)
Supplier: VWR Chemicals
Description: Turbidity standards in kit form or alternatively as formazine stock solutions which allows users to create their own defined standards.
UOM: 1 * 1 KIT
MSDS
Supplier: THERMO FISHER DIAGNOSTICS GMBH
Description: These McFarland equivalence turbidity standard is packed in plastic case with a visual comparision card. Tube size of 15×103 mm.
Catalog Number: (708-0006)
Supplier: Hach
Description: Turbidity Standard, Stablcal® stabilized formazine standard, 4000 NTU, Type: Formazine, 5× 100 ml
UOM: 1 * 1 KIT
Supplier: Hach
Description: Stablcal® turbidity standards for all turbidimeters. USEPA accepted for calibration purposes. Stablcal® stabilised formazine turbidity standards are t...
Supplier: Hach
Description: Gelex secondary standards kit for 2100P and 2100Q series portable turbidimeters (includes 1 sealed vial each for 0 to 10 NTU, 10 to 100 NTU, 100 to 10...
Supplier: Hach
Description: Stablcal® calibration standards kit, for 2100 series portable turbidimeters. USEPA accepted for calibration purposes. Stablcal® stabilised formazin tu...
Supplier: Hach
Description: Turbidity Standard, Stablcal® Turbidity Standards Calibration Kit, < 0,1 NTU, 20 NTU, 200 NTU, 1000 NTU, 4000 NTU, 7500 NTU, Type: Formazine, 6× 100 ml
Catalog Number: (HACHLCW813)
Supplier: Hach
Description: Turbidity Standard, 4000 FNU, Glass bottle
UOM: 1 * 100 mL
Supplier: Hach
Description: Turbidity Standard, Stablcal® Turbidity Standards Calibration Kit, < 0,1 NTU, 20 NTU, 200 NTU, 1000 NTU, 4000 NTU, Type: Formazine, 5× sealed vial
Supplier: Hach
Description: Stablcal Stabilised Formazin Turbidity Standards are true formazin dilutions developed for use in any turbidimeter.
Catalog Number: (HACH2714600)
Supplier: Hach
Description: Stablcal® verification kit containing 0.10 NTU, 0.30 NTU, 0.50 NTU, 1.0 NTU standards (100 ml each), sililcone oil, oiling cloth and instructions is u...
UOM: 1 * 1 KIT
Catalog Number: (REGECRS-40-500)
Supplier: REAGECON
Description: Turbidity standards for ratio and non-ratio instruments composed of suspended polymer microspheres.
UOM: 1 * 500 mL
Supplier: REAGECON
Description: Turbidity standards for ratio and non-ratio instruments composed of suspended polymer microspheres.
Catalog Number: (PROOTURB1000-500ML)
Supplier: LGC Standards PROMOCHEM
Description: Turbidity Standard, Turbidity calibration standard - formazin, 1000 NTU, Type: Formazine
UOM: 1 * 500 mL
Supplier: Thermo Orion
Description: Turbidity Standard, Primary turbidity standards, polymer based, EPA- approved, 0,02 NTU, 20 NTU, 100 NTU, 800 NTU, Type: Polymer suspension, 4 vials
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 18
no targeter for Bottom