You Searched For: Laboratory Reactor Systems
5  results were found
SearchResultCount:"5"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: KINEMATICA
Description: REACTRON® laboratory systems consist of process vessels (single and multiple wall, standard and customer-specific designs), stirring systems for macro...
Supplier: IKA
Description: EasySyn is the reactor system for organic or aqueous syntheses, and the only reactor system for which you can get all the complementary components fro...
Catalog Number: (IKAA20013293)
Supplier: IKA
Description: Modular laboratory reactor for the optimisation and reproduction of various chemical reactions, mixing and homogenisation processes on a lab scale. Th...
UOM: 1 * 1 items
Catalog Number: (IKAA20013291)
Supplier: IKA
Description: Modular laboratory reactor for the optimisation and reproduction of various chemical reactions, mixing and homogenisation processes on a lab scale. Th...
UOM: 1 * 1 items
Catalog Number: (JUCH10101.12)
Supplier: JUCHHEIM Laborgerate GmbH
Description: Laboratory reactor system, 220 °C, 6 bar, complete, includes 2 l glass vessel, 316 stainless steel lid fitted with stirrer head and heating jacket, pressure gauge, excess pressure valve, thermometer pocket, B29 adaptor with pressure tight cap, inlet with plug valve, tripod and a safety cage
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 5 of 5
no targeter for Bottom