Flame-retardant shirts, long sleeved, Devold®
Supplier: Fristads Kansas Group
Devold®
KWIN100621940SEA 0 EUR
KWIN100621940S KWIN100621940XL KWIN1006219402XL KWIN100621940M KWIN100621940L
Flame-retardant shirts, long sleeved, Devold®
Workwear
Long-sleeved shirt made of 52% Lenzing FR® and 48% wool. The two-layer fabric consists of a wool layer on the outside and a layer of Lenzing® FR on the inside. The material transports moisture and keeps the body dry and warm.
  • Lengthened back
  • Also suitable for usage as underwear
Weight: 230 g/m²
Certifications: EN ISO 11612 (A1 B1 C1), EN 61482-1-2 class 1