Reusable half masks, 6300 series
Supplier: 3M
3M
111-0810EA 20.7 EUR
111-0810 111-0812 111-0811
Reusable half masks, 6300 series
Respirators Reusable Respirators Half Face Respirators
This low-maintenance half mask impresses with its easy handling and high wearing comfort. The bayonet click connection enables the filter to be changed quickly and easily.
  • Extremely light mask body for fatigue-free work
  • Compact design allows unrestricted view
  • Double filter system for reduced breathing resistance and good weight distribution
  • Patented double harness for high wearing comfort
Certifications: EN 140