Disposable particle filtering respirators
Supplier: 3M
3M
111-0827EA 0 EUR
111-0827 111-0826 111-0829 111-0825
Disposable particle filtering respirators
Respirators Disposable Respirators Particulate Respirators
The three-part design and the extra soft fleece on the inside of the mask ensure maximum comfort even when worn for a long time.
  • Use limit: up to 4 times the limit for particles
  • Straps: color yellow
  • Folds flat for easy portability
  • Hygienic individual packaging
  • Integrated nose bridge with sweat-absorbing padding, ideal for spectacle wearers
For single use, FFP1 NR D.