Glass dialysis weights
Supplier: Spectrum
SPEC132740EA 0 EUR
SPEC132740
Glass dialysis weights
These can be inserted into any membrane type of tubing to help maintain vertical orientation during dialysis.
The quantity of weights required depends on the tubing length, flat-width and amount of head-space. They are inert and non-binding.