Coomassie brilliant blue G-250, Ultra Pure Grade

Supplier: VWR Chemicals

Synonyms: C.I. 42655

0615-10GEA 57.8 EUR
0615-10G 0615-25G 0615-50G
Coomassie brilliant blue G-250, Ultra Pure Grade
Coomassie brilliant blue G-250

Coomassie Brilliant Blue is a common stain used for the detection of protein bands following electrophoresis.

Formula: C₄₇H₄₈N₃NaO₇S₂
MW: 854,04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00078482
CAS Number: 6104-58-1

Order Now

Specification Test Results

Em (595nm, Water) >= 36300
Protein Staining PASS / Fail

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR