AMREE116
Supplier: Avantor
E116-100MLEA 81.8 EUR
E116-100ML
AMREE116
This BCIP-NBT solution produces a dark blue insoluble end product.
Designed for immunohistochemistry and Northern and Southern hybridisations.