You Searched For: Additives and Supplements for Microbiology
22  results were found
SearchResultCount:"22"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Chemicals
Description: These standard supplements can be used with several culture media to prepare the complete culture media for your specific purpose. Most of these suppl...
Supplier: VWR Chemicals
Description: The isolation of microorganisms often requires the use of specific supplements. The supplements presented here are available in vials and bottles, and...
Supplier: THERMO FISHER DIAGNOSTICS GMBH
Description: MUG supplement, Application: E.coli, Freeze-dried
Supplier: Apollo Scientific
Description: Dehydrated infusion of potato. Plant tissue culture tested.
Supplier: LIOFILCHEM S.R.L.
Description: Yersinia ITC supplement, Application: Additive
Supplier: THERMO FISHER DIAGNOSTICS GMBH
Description: Potassium Lactate 50% , 1 ml, Application: Yeast, Freeze-dried
Catalog Number: (CR102)
Supplier: CHROMAGAR
Description: Chromagar Acinetobacter MDR supplement, Application: Acinetobacter
UOM: 1 * 1 items
Supplier: THERMO FISHER DIAGNOSTICS GMBH
Description: Coagulase Plasma lyophilised, Application: Blood / Serum
Supplier: THERMO FISHER DIAGNOSTICS GMBH
Description: Sheep Blood Defibrinated, Application: Blood/Serum, Freeze-dried
Supplier: BALIS
Description: Additive for Cereus selective agar base acc. to Mossel.
Catalog Number: (USBID9540-05)
Supplier: US Biological
Description: Drop-out mixes are used as a supplement to Drop-out base for making synthetic, complete media formulations in accordance with methods in yeast genetic...
UOM: 1 * 100 g
Catalog Number: (MDTC25-022-CI)
Supplier: Corning
Description: Trace Elements B Solution has the same composition as Cleveland's Trace Element I.
UOM: 1 * 100 mL
Catalog Number: (USBIA0940-500)
Supplier: US Biological
Description: Gelidium based Drosophila agar is an inexpensive growth medium especially suited to the growth and nutrient requirements of Drosophila w...
UOM: 1 * 500 g
Catalog Number: (MDTC25-023-CI)
Supplier: Corning
Description: Trace Elements C Solution has the same composition as Cleveland's Trace Elements I, and contains the trace elements found in the serum-free medium MCD...
UOM: 1 * 100 mL
Supplier: MP Biomedicals
Description: Soy hydrolysate is an enzymatic (papain) digest of soybean meal. It is a plant source peptone. Soy hydrolysate is useful in media where high clarity i...
Catalog Number: (SIALM0553-1VL)
Supplier: Merck
Description: These supplements are ideal for cultivation of Mycobacteria species.
UOM: 1 * 1 Each
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 22
no targeter for Bottom