You Searched For: Base Materials for Microbiology
19  results were found
SearchResultCount:"19"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Chemicals
Description: The ingredients used to prepare culture media are very important, because ultimately the results depend on the quality of these compounds. Raw materia...
Supplier: VWR Chemicals
Description: VWR® is a manufacturer and supplier of high quality biochemicals and reagents for molecular biology, life sciences, proteomics, clinical and histology...
Supplier: VWR Chemicals
Description: Yeast Nitrogen Base (YNB) is a base medium for preparation of minimal and synthetic defined yeast media.
Supplier: THERMO FISHER DIAGNOSTICS GMBH
Description: Yeast extract, powder, 500 g
Supplier: MP Biomedicals
Description: Casamino acids is supplemented to microbiological growth media.
Supplier: LIOFILCHEM S.R.L.
Description: Proteose peptone, 500 g
Supplier: PanReac AppliChem
Description: Components of many microbiological growth media.
Supplier: Thermo Scientific
Description: Yeast extract, 100 g
Supplier: Merck
Description: Yeast extract, 5000 g
Supplier: MP Biomedicals
Description: Yeast hydrolysate enzymatic is an autolyzed yeast extract produced from 100% brewers yeast. It is a concentrated source of soluble protein components,...
Supplier: MP Biomedicals
Description: Beef extract is a water soluble powder, standardized for use in microbiological media.
Supplier: G-Biosciences
Description: Tryptone is an assortment of peptides generated by the tryptic digestion of casein. It is commonly used in microbiology to produce Lysogeny broth for ...
Supplier: MP Biomedicals
Description: Soluble in water (100 mg/ml - clear to slightly hazy solution).
Supplier: HIMEDIA
Description: Peptone, meat Type P, 500 g
Catalog Number: (ICNA0210330401)
Supplier: MP Biomedicals
Description: Yeast Hydrolysate Enzymatic is an autolysed yeast extract produced from 100% brewers yeast. It is a concentrated source of soluble protein components,...
UOM: 1 * 0,45 kg
Supplier: LABORATORIOS CONDA, S.A
Description: Base materials, used in mcrobiology and biochemistry.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 19
no targeter for Bottom