Safety cabinets with wing doors or drawers, Type 30, BASIC LINE
Supplier: Düperthal
DUEH23-201260-402EA 2165.97 EUR
DUEH23-201260-402 DUEH23-201260-401 DUEH23-200660-401 DUEH23-200660-401L DUEH23-060650-401
Safety cabinets with wing doors or drawers, Type 30, BASIC LINE
Safety cabinets with shelves for the storage of hazardous substances in working spaces.
  • High quality mark of approval for upper level style, user-friendliness and increased lifetime
  • Complete cabinet unit made from B1 material
  • Ventilation and extraction at every level
  • Ergonomic lock positioning at handle and integrated lock cylinder in the easy-moving doors
  • Smoothly and quiet closing of the doors by damped bump stops
  • Doors with standard integrated arrest system
Standard equipotential bonding screw on the roof of the cabinet for connecting to the earthing (preventing ignition hazards). The interior fittings are conductively connected with the equipotential bonding screw. In case of fire, the thermocouple initiates the closing of the door automatically.
Adjustable bases with extra-large bearing surface for easy alignment and level adjustment, from the inside and outside. The cabinet is accessible as the design of the base enables easy transport with a hand pallet truck. Base set back to prevent people from hitting their feet on it.
Under bench cabinets are easy moving, single-handed operation of the doors and drawers.
Basic cabinet shelf WxDxH: 515×458×35 mm
Basic XL cabinet shelf WxDxH:1115×458×35 mm
Underbench cabinet shelf WxDxH: 450×350×35 mm
Certifications: Classification Type 30 in accordance with DIN EN 14470-1, TRbF 20 Annex L, TÜV-tested according to DIN EN 14727 (laboratory furniture standard), Type testing according to DIN EN 14470-1, GS mark.