You Searched For: Fittings and Connectors
984  results were found
SearchResultCount:"984"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: Avantor Fluid Handling
Description: Fittings connect in a snap.
Supplier: Avantor Fluid Handling
Description: No more struggling—just press thumb latch.
Supplier: Avantor Fluid Handling
Description: Fittings connect in a snap.
Sale
Supplier: Avantor Fluid Handling
Description: Certificate of compliance included!
Supplier: Avantor Fluid Handling
Description: No tools needed for assembly.
Supplier: Avantor Fluid Handling
Description: Create a secure connection in your sanitary fitting system.
Certificates New Product
Supplier: Avantor Fluid Handling
Description: Meets FDA requirements and 3A sanitary standards.
Catalog Number: (F0001)
Supplier: Avantor
Description: Avantor® ACE®, Replacement ferrules, PEEK
UOM: 1 * 10 items
Supplier: Avantor Fluid Handling
Description: Maintain a permanent connection.
Supplier: Avantor Fluid Handling
Description: Quick and easy assembly without tools.
Supplier: Avantor Fluid Handling
Description: No tools needed for assembly.
Catalog Number: (MFLX31830-74)
Supplier: Avantor Fluid Handling
Description: Convert NPT-threaded systems to BSPP threads—or visa versa—with these stainless steel fittings.
UOM: 1 * 1 items
Supplier: Avantor Fluid Handling
Description: Maintain sanitary conditions.
Supplier: Avantor Fluid Handling
Description: Best for low-pressure applications.
Catalog Number: (HICHHI-454)
Supplier: Avantor
Description: These static mixing tee are ideal for microbore or analytical gradient HPLC. It has a low swept volume (2,2 µl) and is designed for flow rates of 0,5 ...
UOM: 1 * 1 items
Supplier: Avantor Fluid Handling
Description: No tools needed for assembly.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 984
no targeter for Bottom