You Searched For: Nuts
9  results were found
SearchResultCount:"9"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: Bohlender
Description: PTFE, white.
Supplier: COWIE
Description: Universal replacement lock nut for 6 mm shafts
Supplier: COWIE
Description: PTFE, white.
Catalog Number: (LAUDHKM032)
Supplier: LAUDA
Description: Nut, M16×1
UOM: 1 * 1 items
Supplier: BÃœRKLE
Description: HDPE, white.
Catalog Number: (REST24896)
Supplier: Restek
Description: These nuts are ideal for use with Thermo Scientific TRACE GCs.
UOM: 1 * 2 items
Catalog Number: (REST22348)
Supplier: Restek
Description: This hot swap capillary column nut allows to change capillary column while the injector temp is still hot without downtime waiting for injector parts ...
UOM: 1 * 1 items
Catalog Number: (REST22078)
Supplier: Restek
Description: Direct replacement reducing nut, high-quality stainless steel, for Agilent 5890/6850/6890 GC's
UOM: 1 * 1 items
Catalog Number: (HEWL5062-8518)
Supplier: Agilent
Description: Male nut, plastic
UOM: 1 * 4 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 9 of 9
no targeter for Bottom