Universal high-temperature tube furnaces, RHTC
NABERC282O5AEA 0 EUR
NABERC282O5A NABERC282L5A NABERC182O5A NABERC382L5A NABERC182L5A NABERC382O5A
Universal high-temperature tube furnaces, RHTC
These compact tube furnaces with SiC rod heating and integrated switchgear with controller can be used universally for many processes. They represent an inexpensive variant in the high-temperature range. The standard mounting options for accessories make them flexible in use for a wide range of applications. The SiC heating elements arranged parallel to the working tube provide for an excellent temperature uniformity.
The furnace is equipped with standard controller B 510 with touch screen display, featuring 5 freely adjustable programs and 4 segments and 2 extra functions. These controllers are PID microprocessor controlled with a self-diagnostic system and integrated process documentation. The optional controller C 550 has 10 programs with 20 segments each.
Caution: Maximum continuous operating temperature should be 50 to 100 °C below the 'Max. temperature' of the model. Values outside the tube. Difference to temperature inside the tube up to +50 K. External dimensions vary when furnace is equipped with additional equipment.