You Searched For: Circulator Bath Fluids
15  results were found
SearchResultCount:"15"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Chemicals
Description: A range of silicone fluids recommended for use in baths. Low in toxicity, inert and long lasting due to their low volatility.
Supplier: VWR Chemicals
Description: Silicone Oils 47 are polydimethylsiloxane oils, consisting of linear molecular chains of varying lengths whose groups comprise alternating silicone an...
Supplier: JULABO GmbH
Description: Bath fluids have been carefully selected and proven in long-term service. They are suitable for use with heating and refrigerated circulators whenever...
Supplier: Heidolph Instruments GmbH & Co.KG
Description: Non-flammable thermal fluid for heating and low-temperature range. Thermal fluid for Hei-CHILL chillers.
Supplier: Thermo Scientific
Description: MDL No.: MFCD00147787
Density: 0,86
Melting point: –6 °C
Flash point: 230 °C
Appearance: Clear light yellow oily liquid
Stora...
Supplier: LAUDA
Description: Allow highly accurate thermostating, even at extreme temperatures.
Supplier: LAUDA
Description: Open systems are atmospherically open, as semi-open systems are referred to open baths with lids. The thermally active heat transfer medium in closed ...
Supplier: LAUDA
Description: Heat transfer liquid for use in systems with cold liquid overlay.
New Product
Supplier: IKA
Description: Type: Heating fluid, Pk: 10 kg, Temperature range: +20...+200 °C, Colour: Clear
Supplier: LAUDA
Description: These transfer fluids allow highly accurate thermostating, even at extreme temperatures.
Supplier: LAUDA
Description: These transfer fluids allow highly accurate thermostating, even at extreme temperatures.
Supplier: DOW CORNING
Description: A range of silicone fluids recommended for use in baths. Low in toxicity, inert and long lasting due to their low volatility.
Supplier: HUBER
Description: Thermal fluids with a wide range of operating temperatures and good thrmodynamic properties for use in circulators and heating systems where water is ...
Supplier: Merck
Description: Silicone fluid, Pk: 5 L, Temperature range: 20…60 °C
Supplier: THERMO ELECTRON LED GMBH
Description: These carefully selected and proven heat transfer liquids have been designed to reduce health hazards and minimise unpleasant odours.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 15 of 15
no targeter for Bottom