You Searched For: Electrode Storage Solutions
19  results were found
SearchResultCount:"19"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Chemicals
Description: For full information concerning health and safety data please see the website.
Catalog Number: (ORIO951213)
Supplier: Thermo Orion
Description: Take more accurate and reproducible ISE measurements with Thermo Scientificâ„¢ Orionâ„¢ ISE Ionic Strength Adjustors (ISA) and special reagents. Ionic str...
UOM: 1 * 1 items
Catalog Number: (662-9223)
Supplier: HAMILTON BONADUZ
Description: Hamilton storage solution is recommended to increase sensor longevity when not in use. The liquid solution is acid buffered to help with sensor respon...
UOM: 1 * 500 mL
MSDS
Catalog Number: (HACHC20C370)
Supplier: Hach
Description: Electrode cleaning solution to remove proteins residues organic contaminations/deposits such as food stuff from pH sensing element.
UOM: 1 * 250 mL
Catalog Number: (HACH2965349)
Supplier: Hach
Description: Used to condition ORP electrode surfaces for use in reducing sample matrices.
UOM: 1 * 1 items
Catalog Number: (705-1031)
Supplier: Hach
Description: Electrode cleaning solution, for cleaning all pH, ORP and sodium ion selective eletrodes, 500 ml bottle
UOM: 1 * 500 mL
Supplier: Mettler - Toledo
Description: Pepsin-HCl for cleaning junctions with protein contamination, 250 ml
Catalog Number: (HACH2657364)
Supplier: Hach
Description: pH electrode storage solution, powder pillows (each pillow makes 25 ml of storage solution)
UOM: 1 * 20 items
Catalog Number: (705-1030)
Supplier: Hach
Description: Potassium chloride (KCl) 3 M
UOM: 1 * 500 mL
Catalog Number: (HACHS16M001)
Supplier: Hach
Description: Mild cleaning solution for electrodes. Ideal for use after measurements in aqueous solutions.
UOM: 1 * 250 mL
Catalog Number: (HACH2541249)
Supplier: Hach
Description: For storage of ammonia ion selective electrodes.
UOM: 1 * 500 mL
Catalog Number: (HACHS16M002)
Supplier: Hach
Description: This extra strong electrode cleaning solution is ideal for use when stronger cleaning agents are required, for example when the solutions contain prot...
UOM: 1 * 250 mL
Catalog Number: (662-1548)
Supplier: Thermo Orion
Description: Restore proper electrode performance and prolong electrode life using Thermo Scientificâ„¢ Orionâ„¢ pH electrode storage solutions.
UOM: 1 * 475 mL
MSDS
Supplier: Thermo Orion
Description: Solutions for storage and cleaning that prolong the working life of pH electrodes. Cleaning solutions are identified by the letters A, B, C, and D; ea...
Catalog Number: (HAMI238290)
Supplier: HAMILTON BONADUZ
Description: A two-part cleaning solution for all glass membrane pH sensors. pH Cleaning solution A contains sodium hydroxide (<3%), pH cleaning solution B con...
UOM: 1 * 1 SET
MSDS
Supplier: Hanna
Description: A variety of cleaning solutions for electrodes.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 19
no targeter for Bottom