Eco stopcock plugs, with taper 1:5
Supplier: Bohlender
BOLA
BOHLJ1205-13EA 12.5 EUR
BOHLJ1205-13 BOHLJ1225-10 BOHLJ1205-28 BOHLJ1205-18 BOHLJ1205-19 BOHLJ1225-15 BOHLJ1225-05 BOHLJ1225-35 BOHLJ1225-25
Eco stopcock plugs, with taper 1:5
Stopcocks
Made of PFTE, white, with grip and nut of PP, red.
  • Universal chemical resistance
  • Temperature resistance: from -10 °C to +120 °C
  • Taper: 1:5
  • With or without bore
  • Washer included