Corning® Human collagen III (from placenta)
Supplier: Corning
734-0104EA 397.92 EUR
BDAA354244
Corning® Human collagen III (from placenta)
Collagen III is found in several stromal connective tissues including the dermis of young organisms, human skin, and cornea. It can be used as a thin coating on tissue culture surfaces to promote cell attachment and to modulate cell behaviour.
Quality controlled by SDS-PAGE and tested and found negative for bacteria, fungi, and mycoplasma, Source material tested for hepatitis B antigen and HIV-1 antibody