Bottle-top dispenser, Volupette
612-3873EA 268 EUR
JENC263-027
Bottle-top dispenser, Volupette
Variable volume, autoclavable.
  • Wetted parts autoclavable at +121 °C
  • Easily removable PTFE piston for smooth action and easy cleaning
  • Safety pedestal valve allows the Volupette to be turned off to prevent accidental delivery, whilst ensuring it is drip-free
  • Easy to disassemble glass barrel for thorough cleaning
  • With standard 30 mm bottle neck fitting
The Volupette is a variable bottle-top dispenser that can be used as a manually primed unit or, once fitted with the supplied spring, converted to a fully self-priming dispenser. The dispense volume is easily adjusted by rotating the top knob to move the indicator to the desired setting.
Ordering information: Supplied with three bottle neck adapters (38, 40 and 45 mm), spring, calibration certificate and instruction manual. Please note that the Volupette is not supplied with a reservoir.