Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
cloning-reagents
https://ru.vwr.com/stibo/smallweb/std.lang.all/50/44/18945044.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"37"
  List View Grid View Easy View (new)
Sort by:
Supplier:  Quantabio
Description:   RepliQa™ HiFi Assembly Mix is a one-step, isothermal reaction for the cloning of single and multi-insert assemblies eliminating the need of restrictio...
Catalog Number: (27-4563-01)
Supplier:  GE Healthcare - Life Sciences
Description:   Calf thymus DNA sheared by sonication and provided lyophilised in a 100 OD pack size.
UOM:  1 * 1 items
Catalog Number: (ROCKMB-103-1000)
Supplier:  Rockland Immunochemicals
Description:   DNA from salmon is commonly used in the preparation of pre-hybridisation and hybridisation solutions. These prepared solutions are used to block the n...
UOM:  1 * 1 g
Catalog Number: (27-4111-01)
Supplier:  GE Healthcare - Life Sciences
Description:   Lambda DNA (1 ml pack), provided at concentration of 500 µg/ml.
UOM:  1 * 500 µG
Supplier:  SGI - DNA Synthetic Genomics
Description:   The Gibson Assembly® Ultra Kit provides a robust and efficient method to seamlessly construct synthetic genes, genetic pathways, as well as entire gen...
Supplier:  SGI - DNA Synthetic Genomics
Description:   The Gibson Assembly® Method is a well-established assembly reaction that can be leveraged to join multiple, mutagenised DNA fragments with overlapping...
Catalog Number: (27-1524-01)
Supplier:  GE Healthcare - Life Sciences
Description:   M13 phage for the GST expression system.
UOM:  1 * 100 µl
Catalog Number: (ROCKMB-104-0500)
Supplier:  Rockland Immunochemicals
Description:   Lambda phage consists of a virus particle including a head (also known as a capsid), a tail and tail fibres. The head contains the phage's double-stra...
UOM:  1 * 500 µG
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 8  of 37
  1  2  3  4  5  Next