Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
    • In Stock (9)
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
Printers
https://ru.vwr.com/stibo/smallweb/std.lang.all/96/80/18989680.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"44"
  List View Grid View Easy View (new)
Sort by:
Supplier:  VWR Collection
Description:   Thermal printer TLP-50 for VWR LA, LP, SM balances and MB moisture analysers, EU-plug + UK-plug, UK/EU-plugs, Thermal
Catalog Number: (720-2890)
Supplier:  VWR Collection
Description:   The Signature slide printer can significantly increase laborator efficiency while helping to reduce the risk of incorrect sample identification. It pr...
UOM:  1 * 1 items
Catalog Number: (611-3623)
Supplier:  VWR Collection
Description:   The statistical printer is a bidirectional thermal printer that can be used with laboratory balances. It can print weight values and, when requested, ...
UOM:  1 * 1 items
Supplier:  BRADY & CO LTD, W. H.
Description:   The BradyJet J5000™ colour label printer produces safety signs & labels in colour within a few seconds. BradyJet J5000 quickly prints safety signs, le...
Supplier:  BRADY & CO LTD, W. H.
Description:   The BMP™41 portable label printer creates die-cut or continuous labels anywhere and at anytime. It can produce up to 25 mm wide labels which can be us...
Supplier:  BRADY & CO LTD, W. H.
Description:   Mobile, thermal transfer, label printer that can print on more than 30 different types of of label including heat shrink sleeves, die-cut labels or in...
Supplier:  BRADY & CO LTD, W. H.
Description:   Brady IP™ printer is a fully integrated, easy-to-use printer, material, and software system. Smart Cell technology allows the printer, materials, and ...
Supplier:  CHROMACOL
Description:   From sample entry to final reports, everything an analytical lab does revolves around ensuring accurate data and reliable results. Chromatography vial...
UOM:  1 * 1 items
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 8  of 44
  1  2  3  4  5  6  Next