You Searched For: Dispersing Tools
11  results were found
SearchResultCount:"11"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: Choice of aggregates for a wide range of laboratory applications from 5 ml up to 20 L. The VWR® Aggregates solve the standard problems that are faced ...
Sale
Catalog Number: (710-0953)
Supplier: VWR Collection
Description: Dispersing Element, Johnny sealing ring for blades, autoclavable, for use with Steri-Blostev 12 base fixing plate
UOM: 1 * 1 items
Supplier: KINEMATICA
Description: BIOTRONA® design high turbulence mixing. High-turbulence mixing head with low shear forces and energy inputs. Time saving with improved results compar...
Supplier: KINEMATICA
Description: Designation: EC - EasyClean version for the simplest cleaning or sterilisation (Standard)/2 - rotor/stator with 1 tooth row each.
Supplier: KINEMATICA
Description: Dispersing aggregates available as standard (EC design) and special aggregates (W, B design).
Catalog Number: (431-0205)
Supplier: KINEMATICA
Description: Dispersing element, PT-DA 20/2EC-E192, For unit: PT 4000, Speed: 27 m/s, Stator/rotor Ø: 15 / 20 mm, Volume: 100 - 2500 ml
UOM: 1 * 1 items
Supplier: IKA
Description: Type N: Materials in contact with medium are PTFE and stainless steel AISI 316L, suitable for solvents and abrasive substances, can be sterilis...
Supplier: IKA
Description: Disposable, plastic dispersing elements are ideal for applications where cross-contamination cannot be tolerated. Dispersing elements can be thrown aw...
Supplier: IKA
Description: Dispersing element, D 60, Stator/rotor Ø: -/60 mm, Volume: 2 - 30 l
Supplier: VELP SCIENTIFIC
Description: A wide range of dispersing tools ensures excellent performance for many applications for small samples, sizes such as biological matrices, and large v...
New Product
Catalog Number: (WITGDH.WHG02108)
Supplier: WITEG LABORTECHNIK GMBH
Description: Dispersing Element, Homogeniser tool, HT1008, For: HG-15A, WiseTis®, Stator/rotor Ø: 8,0/6,1 mm, Length: 138 mm, Volume: 1 - 50 ml
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 11 of 11
no targeter for Bottom