Magnetic separation devices
Supplier: Omega Bio-Tek
VWR®
730-1400EA 334 EUR
730-1400
Magnetic separation devices
Racks Magnetic Separation Racks
A range of magnetic devices for separation of proteins or peptides 'bound' to magnetic particles or beads. Magnetic separation is a simple, scaleable and gentle technique that is suitable for crude samples, even those containing particulates.
  • Devices available for 96-well plates and microtubes
  • Simple and effective immobilastion of particles whilst the buffer/elutant is exchanged