You Searched For: Sieves for Mills
17  results were found
SearchResultCount:"17"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: KINEMATICA
Description: Sieve, for mill, POLYMIX® system PX-MFC 90 D, Insert sieve, stainless steel 1.4034, Mesh size: 6,00 mm
Supplier: Retsch GmbH
Description: Sieve, for mills, Steel, Ring sieves, trapezoid holes, Mesh size: 0,25 mm, Ø: 70 mm
Supplier: Retsch GmbH
Description: Sieve, for mills, Stainless steel, Sieves 180°, round holes, Mesh size: 4,00 mm
Supplier: Retsch GmbH
Description: Sieve, for mills, Stainless steel, Bottom sieves, square holes, Mesh size: 6,00 mm
Supplier: Retsch GmbH
Description: Sieve, for mills, Titanium, Ring sieve, round holes, with reinforced rim, Mesh size: 4,00 mm
Supplier: Retsch GmbH
Description: Sieve, for mills, Stainless steel, Ring sieve, trapezoid holes, with strengthened edges, recommended for heavy-duty usage, Mesh size: 1,00 mm
Supplier: Retsch GmbH
Description: Sieve, for mills, Stainless steel, Distance sieve, trapezoid holes, Mesh size: 2,00 mm
Supplier: Retsch GmbH
Description: Sieve, for mills, Stainless steel, Ring sieve, trapezoid holes, with strengthened edges, Mesh size: 0,75 mm
Supplier: Retsch GmbH
Description: Sieve, for mills, Stainless steel, Bottom sieves, trapezoid holes, Mesh size: 0,50 mm
Supplier: IKA
Description: Sieve, for mills, Stainless steel sieve, 2,00 mm
Supplier: IKA
Description: Interchangeable sieves for insertion into the grinding heads ensure maximum particle size filtering.
New Product
Catalog Number: (412-2354)
Supplier: Retsch GmbH
Description: Sieve, for mills, Stainless steel, Bottom sieves, trapezoid holes, Mesh size: 0,50 mm
UOM: 1 * 1 items
Supplier: Retsch GmbH
Description: Sieve, for mills, Bottom sieve, trapezoid holes, Mesh size: 0,75 mm
Supplier: FRITSCH
Description: Sieve, for mills, Stainless steel, Sieve cassettes, trapezoid perforation, made of stainless steel 316, Mesh size: 1,00 mm
Supplier: FRITSCH
Description: Sieve cassettes determine the fineness of a sample. Sieve cassettes are provided with trapezoidal and square perforations.
Supplier: FRITSCH
Description: Sieve inserts determine the final fineness of a sample. These sieve inserts are available with trapezoidal or square perforations.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 17
no targeter for Bottom