Calculator converter provides fast and accurate answers for 126 types of metric conversions. Simply enter a number, press an arrow, and read the exact metric or U.S. answer with decimal point in place. Raised, oversized keys make the metric converter a joy to use, even for large fingers. It includes an automatic flip open LCD cover that converts to a bench top stand, rubberised side grips for easy handling, and a bright, vivid digit LCD that is easy to read.
- Change lab measurements to U.S. or metric in an instant
- Easy to use and easy to read
- Ultra-lightweight and compact for carrying in a pocket
Functions: Converts length, area, volume, weight, liquid, energy and temperature. Also performs English (Imperial) conversions and all routine calculator functions.