INTERFERin®-HTS is a siRNA transfection reagent especially developed for High Throughput Screening (HTS) applications providing great silencing efficiency, excellent reproducibility and high cell viability with very low amounts of reagent. INTERFERin®-HTS is cost-effective, easy to handle, compatible with serum and antibiotics, and comes with reverse and forward protocols for 96- and 384-well plates. For routine transfections see INTERFERin® siRNA tranfection reagent.
- Great silencing from 10 to 50 nM siRNA
- Designed for automated procedures
- Reproducible at all levels
- Cost effective, using only a small volume of reagent per well
Certifications: Polyplus-transfection® has been an ISO 9001 accredited company since 2002. Since then this certification has been successfully renewed every three years.
Delivery information: 1,5 ml of INTERFERin®-HTS is sufficient to transfect 200 to 300 96-well plates.