Affinity chromatography sorbents, Heparin HyperD® M
Supplier: Pall Laboratory
555-0004EA 6660 EUR
PALL20029-013
Affinity chromatography sorbents, Heparin HyperD® M
High speed, high capacity affinity preparative sorbent for the purification of biological molecules that bind heparin, such as coagulation factors, growth factors, and lipoproteins.
  • High binding capacity and high flow rates
  • Ideal for small- and large-scale applications
Dynamic sorption capacity for Human ATIII (10% breakthrough)*: 25 mg/ml.
Immobilised heparin: 5 to 10 mg/ml of sorbent
* Determined using human ATIII at 72,5 UI/ml in 20 mM Tris-HCl, 0,3 M NaCl, pH 7,4. Elution with 20 mM Tris HCl, 2 M NaCl, pH 7,4 at a flow rate of 600 cm/h, 10 cm bed height.