Delivery reagent, in vivo-jetPEI®-FluoF
Supplier: Polyplus-transfection
205-10GEA 0 EUR
PPLU205-10G
Delivery reagent, in vivo-jetPEI®-FluoF
in vivo-jetPEI®-FluoF delivery reagent is a fluorescein-conjugated linear polyethylenimine derivative (green label) designed to perform biodistribution studies using fluorescent labelling.
  • Excitation at 490 nm; emission at 520 nm
  • Easy to use
Certifications: Polyplus-transfection® has been an ISO 9001 accredited company since 2002. Since then this certification has been successfully renewed every three years.
Delivery information: 10% glucose solution is included. 0,1 ml of in vivo-jetPEI®-FluoF delivery reagent is sufficient to perform up to 20 intravenous injections in mouse with 50 µg of DNA.