FectoFly® DNA transfection reagent is dedicated to the transfection of insect cells for the production of baculovirus and recombinant proteins. FectoFly® provides high level of transgene expression in both adherent and suspension insect cells, such as S2, SF9, SF21 and TN5.
- High protein expression and baculovirus production
- Global transfection solution for insect cells
- High transfection efficiency in adherent and suspension insect cells
- Does not contain any components of animal origin
- Easy to use and compatible with the use of serum and antibiotics
Certifications: Polyplus-transfection® has been an ISO 9001 accredited company since 2002. Since then this certification has been successfully renewed every three years.
Delivery information: 150 mM NaCl solution included. 1 ml of FectoFly® is sufficient to perform 100 to 200 transfections in 6-well plates.